4-Bromo-2-fluorostyrene

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-fluoro-1-vinylbenzene, 4-Bromo-1-ethenyl-2-fluorobenzene

PC7069-1G PC7069-5G
APOSPC7069-1GEA 244 EUR
APOSPC7069-1G APOSPC7069-5G
4-Bromo-2-fluorostyrene
4-Bromo-2-fluorostyrene
Formel: C₈H₆BrF
Molecular Weight: 201.036 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09907860
CAS-Nummer: 627463-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR