Pentafluorophenyl 2-morpholin-4-ylbenzoate 97%
Lieferant: Apollo Scientific
Synonyme: Pentafluorophenyl 2-morpholinobenzoate
PC7100-1G PC7100-250MG
APOSPC7100-1GEA 220 EUR
APOSPC7100-1G APOSPC7100-250MG
Pentafluorophenyl 2-morpholin-4-ylbenzoate 97%
Pentafluorophenyl 2-morpholin-4-ylbenzoate
Formel: C₁₇H₁₂F₅NO₃
Molecular Weight: 373.274 g/mol
Schmelzpunkt: 86.5…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025824
CAS-Nummer: 906352-59-8