1-(4-Bromo-2-fluorobenzyl)pyrrolidine

Lieferant: Apollo Scientific
Gefahr

PC7159-25G PC7159-5G
APOSPC7159-25GEA 277 EUR
APOSPC7159-25G APOSPC7159-5G
1-(4-Bromo-2-fluorobenzyl)pyrrolidine
1-(4-Bromo-2-fluorobenzyl)pyrrolidine
Formel: C₁₁H₁₃BrFN
Molecular Weight: 258,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09749655
CAS-Nummer: 283173-83-1
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR