Hexakis(1H,1H,9H-perfluorononyloxy)phosphazene 95%

Lieferant: Apollo Scientific
Achtung

PC7165-1G PC7165-250MG
APOSPC7165-1GEA 630 EUR
APOSPC7165-1G APOSPC7165-250MG
Hexakis(1H,1H,9H-perfluorononyloxy)phosphazene 95%
Hexakis(1H,1H,9H-perfluorononyloxy)phosphazene
Formel: C₅₄H₁₈F₉₆N₃O₆P₃
Molecular Weight: 2721,505 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784334
CAS-Nummer: 186043-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR