8-Fluoro-4-hydroxy-2-methylquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Fluoro-4-hydroxy-2-methyl-1-azanaphthalene, 8-Fluoro-2-methylquinolin-4-ol, 8-fluoro-2-methyl-4-quinolinol

PC7189-1G PC7189-500MG PC7189-5G
APOSPC7189-1GEA 56.7 EUR
APOSPC7189-1G APOSPC7189-500MG APOSPC7189-5G
8-Fluoro-4-hydroxy-2-methylquinoline
8-Fluoro-4-hydroxy-2-methylquinoline
Formel: C₁₀H₈FNO
Molecular Weight: 177,175 g/mol
Schmelzpunkt: 230…236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00272371
CAS-Nummer: 5288-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR