4-Amino-2,8-bis(trifluoromethyl)quinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,8-Bis(trifluoromethyl)quinolin-4-amine

PC7196-1G PC7196-5G
APOSPC7196-1GEA 297 EUR
APOSPC7196-1G APOSPC7196-5G
4-Amino-2,8-bis(trifluoromethyl)quinoline
4-Amino-2,8-bis(trifluoromethyl)quinoline
Formel: C₁₁H₆F₆N₂
Molecular Weight: 280.169 g/mol
Schmelzpunkt: 157…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08669716
CAS-Nummer: 917561-99-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR