1-[3-(4-Fluorophenoxy)prop-1-yl]piperazine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Fluoro-4-[3-(piperazin-1-yl)propoxy]benzene

PC7237-5G PC7237-1G
APOSPC7237-5GEA 522 EUR
APOSPC7237-5G APOSPC7237-1G
1-[3-(4-Fluorophenoxy)prop-1-yl]piperazine 97%
1-[3-(4-Fluorophenoxy)prop-1-yl]piperazine
Formel: C₁₃H₁₉FN₂O
Molecular Weight: 238.301 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094420
CAS-Nummer: 91940-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR