2-Bromo-5-fluorostyrene

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Bromo-2-ethenyl-4-fluorobenzene, 1-Bromo-4-fluoro-2-vinylbenzene

PC7261-1G PC7261-250MG
APOSPC7261-1GEA 140 EUR
APOSPC7261-1G APOSPC7261-250MG
2-Bromo-5-fluorostyrene
2-Bromo-5-fluorostyrene
Formel: C₈H₆BrF
Molecular Weight: 201.036 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09749661
CAS-Nummer: 828267-47-6
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR