5-Bromo-2-fluoro-4-methylbenzeneboronic acid 98%
Lieferant: Apollo Scientific
Synonyme:
5-Bromo-2-fluoro-4-methylphenylboronic acid
PC7275-1G
PC7275-5G
APOSPC7275-1GEA
92.8
EUR
APOSPC7275-1G
APOSPC7275-5G
5-Bromo-2-fluoro-4-methylbenzeneboronic acid 98%
5-Bromo-2-fluoro-4-methylbenzeneboronic acid
Formel:
C₇H₇BBrFO₂ Molecular Weight: 232.843 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258744 CAS-Nummer: 957061-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...