6-Bromo-2-fluoro-3-methylbenzeneboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(6-Bromo-2-fluoro-3-methylphenyl)boronic acid, 3-Borono-4-bromo-2-fluorotoluene, 6-Bromo-2-fluoro-3-methylphenylboronic acid
PC7276-1G
PC7276-25G
PC7276-5G
APOSPC7276-1GEA
65.5
EUR
APOSPC7276-1G
APOSPC7276-25G
APOSPC7276-5G
6-Bromo-2-fluoro-3-methylbenzeneboronic acid 98%
6-Bromo-2-fluoro-3-methylbenzeneboronic acid
Formel:
C₇H₇BBrFO₂ Molecular Weight: 232,843 g/mol Schmelzpunkt: 136…139 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09258741 CAS-Nummer: 957061-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...