1H,1H,10H,10H-Hexadecafluorodecane-1,10-diyl bis(2-propylpentanoate)

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluorodecane-1,10-diyl bis(2-propylpentanoate)

PC7286-25G PC7286-5G
APOSPC7286-25GEA 102 EUR
APOSPC7286-25G APOSPC7286-5G
1H,1H,10H,10H-Hexadecafluorodecane-1,10-diyl bis(2-propylpentanoate)
1H,1H,10H,10H-Hexadecafluorodecane-1,10-diyl bis(2-propylpentanoate)
Formel: C₂₆H₃₄F₁₆O₄
Molecular Weight: 714,52 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059546
CAS-Nummer: 544446-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR