4-Chlor-3-fluorbenzoylchlorid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC7293-100G
PC7293-25G
PC7293-5G
APOSPC7293-100GEA
1020
EUR
APOSPC7293-100G
APOSPC7293-25G
APOSPC7293-5G
4-Chlor-3-fluorbenzoylchlorid 97%
4-Chlor-3-fluorbenzoylchlorid
Formel:
C₇H₃Cl₂FO Lagertemperatur: Raumtemperatur |
MDL:
MFCD02091532 CAS-Nummer: 160658-68-4 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...