4,4,4-Trifluorobutanamide 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,4,4-Trifluorobutyramide
PC7347-1G
PC7347-5G
APOSPC7347-1GEA
271
EUR
APOSPC7347-1G
APOSPC7347-5G
4,4,4-Trifluorobutanamide 97%
4,4,4-Trifluorobutanamide
Formel:
C₄H₆F₃NO Molecular Weight: 141,092 g/mol Schmelzpunkt: 137…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00526200 CAS-Nummer: 461-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...