2-Bromo-5-fluorobenzylamine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
PC7384-1G
PC7384-25G
PC7384-5G
APOSPC7384-1GEA
39.3
EUR
APOSPC7384-1G
APOSPC7384-25G
APOSPC7384-5G
2-Bromo-5-fluorobenzylamine hydrochloride
2-Bromo-5-fluorobenzylamine hydrochloride
Formel:
C₇H₇BrFN·ClH Molecular Weight: 240,501 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142877 CAS-Nummer: 202865-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...