3-Chloro-2,4-difluoroiodobenzene 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-1,3-difluoro-4-iodobenzene

PC7421-1G PC7421-25G PC7421-5G
APOSPC7421-1GEA 82.8 EUR
APOSPC7421-1G APOSPC7421-25G APOSPC7421-5G
3-Chloro-2,4-difluoroiodobenzene 97%
3-Chloro-2,4-difluoroiodobenzene
Formel: C₆H₂ClF₂I
Molecular Weight: 274,434 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00142890
CAS-Nummer: 202925-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR