3-Acetyl-5-bromo-2-fluoropyridine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(5-Bromo-2-fluoropyridin-3-yl)ethan-1-one, 1-(5-Bromo-2-fluoropyridin-3-yl)ethanone

PC7516-25G PC7516-5G
APOSPC7516-25GEA 357 EUR
APOSPC7516-25G APOSPC7516-5G
3-Acetyl-5-bromo-2-fluoropyridine 98%
3-Acetyl-5-bromo-2-fluoropyridine
Formel: C₇H₅BrFNO
Molecular Weight: 218,023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12024449
CAS-Nummer: 1111637-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR