4-Bromo-1,1,1,3,3-pentafluorobutane 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,4,4,4-Pentafluorobutyl bromide

PC7567-250MG
APOSPC7567-250MGEA 115 EUR
APOSPC7567-250MG
4-Bromo-1,1,1,3,3-pentafluorobutane 97%
4-Bromo-1,1,1,3,3-pentafluorobutane
Formel: C₄H₄BrF₅
Molecular Weight: 226,971 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08460470
CAS-Nummer: 933600-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR