4-Bromo-2-fluoro-6-methoxybenzeneboronic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-fluoro-6-methoxyphenylboronic acid, (4-Bromo-2-fluoro-6-methoxyphenyl)boronic acid, 2-Borono-5-bromo-3-fluoroanisole

PC7585-1G PC7585-5G
APOSPC7585-1GEA 68.3 EUR
APOSPC7585-1G APOSPC7585-5G
4-Bromo-2-fluoro-6-methoxybenzeneboronic acid 95+%
4-Bromo-2-fluoro-6-methoxybenzeneboronic acid
Formel: C₇H₇BBrFO₃
Molecular Weight: 248,842 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09475882
CAS-Nummer: 957035-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR