3-Acetyl-2-methyl-6-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]ethan-1-one, 3-Ethanoyl-2-methyl-6-(trifluoromethyl)pyridine

PC7652-1G PC7652-5G
APOSPC7652-1GEA 154 EUR
APOSPC7652-1G APOSPC7652-5G
3-Acetyl-2-methyl-6-(trifluoromethyl)pyridine
3-Acetyl-2-methyl-6-(trifluoromethyl)pyridine
Formel: C₉H₈F₃NO
Molecular Weight: 203,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407376
CAS-Nummer: 205582-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR