Hexafluoro-3-(trifluoromethyl)-1-iodobut-1-ene

Lieferant: Apollo Scientific

PC7684-5G PC7684-1G
APOSPC7684-5GEA 294 EUR
APOSPC7684-5G APOSPC7684-1G
Hexafluoro-3-(trifluoromethyl)-1-iodobut-1-ene
Hexafluoro-3-(trifluoromethyl)-1-iodobut-1-ene
Formel: C₅F₉I
Molecular Weight: 357,944 g/mol
Siedepunkt: 92…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075277
CAS-Nummer: 105774-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR