2-Bromo-2'-fluoropropiophenone

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(2-fluorophenyl)propan-1-one

PC7689-1G PC7689-250MG PC7689-5G
APOSPC7689-1GEA 284 EUR
APOSPC7689-1G APOSPC7689-250MG APOSPC7689-5G
2-Bromo-2'-fluoropropiophenone
2-Bromo-2'-fluoropropiophenone
Formel: C₉H₈BrFO
Lagertemperatur: Raumtemperatur
MDL: MFCD09955286
CAS-Nummer: 186036-09-9
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR