2-Bromo-5-fluorophenylacetonitrile 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-fluorobenzyl cyanide, 2-(2-Bromo-5-fluorophenyl)acetonitrile, (2-Bromo-5-fluorophenyl)acetonitrile

PC7746-1G PC7746-25G PC7746-5G
APOSPC7746-1GEA 21.9 EUR
APOSPC7746-1G APOSPC7746-25G APOSPC7746-5G
2-Bromo-5-fluorophenylacetonitrile 99%
2-Bromo-5-fluorophenylacetonitrile
Formel: C₈H₅BrFN
Molecular Weight: 214,034 g/mol
Schmelzpunkt: 70…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094439
CAS-Nummer: 886761-96-2
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR