4-Bromo-5-chloro-2-fluorotoluene 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-chloro-5-fluoro-4-methylbenzene
PC7757-25G
PC7757-100G
APOSPC7757-25GEA
127
EUR
APOSPC7757-25G
APOSPC7757-100G
4-Bromo-5-chloro-2-fluorotoluene 98%
4-Bromo-5-chloro-2-fluorotoluene
Formel:
C₇H₅BrClF Molecular Weight: 223.47 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143269 CAS-Nummer: 201849-17-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...