4-(Trifluoromethyl)pyridine-3-thiocarboxamide 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Trifluoromethyl)nicotinthioamide
PC7766T-1G
PC7766T-5G
APOSPC7766T-1GEA
126
EUR
APOSPC7766T-1G
APOSPC7766T-5G
4-(Trifluoromethyl)pyridine-3-thiocarboxamide 95+%
4-(Trifluoromethyl)pyridine-3-thiocarboxamide
Formel:
C₇H₅F₃N₂S Molecular Weight: 206,188 g/mol Schmelzpunkt: 153…156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00205806 CAS-Nummer: 158063-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...