4-Fluoronaphthalene-1-sulphonyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(Chlorosulphonyl)-4-fluoronaphthalene
PC7792-1G
PC7792-500MG
PC7792-5G
APOSPC7792-1GEA
0
EUR
APOSPC7792-1G
APOSPC7792-500MG
APOSPC7792-5G
4-Fluoronaphthalene-1-sulphonyl chloride 97%
4-Fluoronaphthalene-1-sulphonyl chloride
Formel:
C₁₀H₆ClFO₂S Molecular Weight: 244,67 g/mol Schmelzpunkt: 80…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02091376 CAS-Nummer: 316-69-8 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...