6,6,6-Trifluoro-DL-norleucine 96+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-6,6,6-trifluorohexanoic acid, 2-Amino-6,6,6-trifluorocaproic acid

PC7812-1G PC7812-250MG
APOSPC7812-1GEA 487 EUR
APOSPC7812-1G APOSPC7812-250MG
6,6,6-Trifluoro-DL-norleucine 96+%
6,6,6-Trifluoro-DL-norleucine
Formel: C₆H₁₀F₃NO₂
Molecular Weight: 185,144 g/mol
Schmelzpunkt: 272…274 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00190647
CAS-Nummer: 120200-04-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR