Methyl 2-nitro-4-(trifluoromethyl)benzoate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-nitro-α,α,α-trifluoro-p-toluate, 4-(Methoxycarbonyl)-3-nitrobenzotrifluoride
PC7828-1G
PC7828-25G
APOSPC7828-1GEA
21.9
EUR
APOSPC7828-1G
APOSPC7828-25G
Methyl 2-nitro-4-(trifluoromethyl)benzoate 98%
Methyl 2-nitro-4-(trifluoromethyl)benzoate
Formel:
C₉H₆F₃NO₄ Molecular Weight: 249,143 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08282772 CAS-Nummer: 228418-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...