4,4,4-Trifluoro-3-(trifluoromethyl)butan-1-ol 97%

Lieferant: Apollo Scientific

Synonyme: 1-Hydroxy-4,4,4-trifluoro-3-(trifluoromethyl)butane

PC7854E-1G PC7854E-5G
APOSPC7854E-1GEA 234 EUR
APOSPC7854E-1G APOSPC7854E-5G
4,4,4-Trifluoro-3-(trifluoromethyl)butan-1-ol 97%
4,4,4-Trifluoro-3-(trifluoromethyl)butan-1-ol
Formel: C₅H₆F₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD00236628
CAS-Nummer: 17327-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR