4,4,4-Trifluoro-3-(trifluoromethyl)butanoic acid 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3,3-Bis(trifluoromethyl)propanoic acid, 4,4,4-Trifluoro-3-(trifluoromethyl)butyric acid

PC7854L-1G PC7854L-5G
APOSPC7854L-1GEA 65.5 EUR
APOSPC7854L-1G APOSPC7854L-5G
4,4,4-Trifluoro-3-(trifluoromethyl)butanoic acid 97%
4,4,4-Trifluoro-3-(trifluoromethyl)butanoic acid
Formel: C₅H₄F₆O₂
Molecular Weight: 210,074 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236630
CAS-Nummer: 17327-33-2
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR