Methyl perfluoro-3,6,9-trioxatridecanoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl difluoro{2-[2-(nonafluorobutoxy)-1,1,2,2-tetrafluoroethoxy]-1,1,2,2-tetrafluoroethoxy}acetate

PC7879-25G PC7879-5G
APOSPC7879-25GEA 285 EUR
APOSPC7879-25G APOSPC7879-5G
Methyl perfluoro-3,6,9-trioxatridecanoate
Methyl perfluoro-3,6,9-trioxatridecanoate
Formel: C₁₁H₃F₁₉O₅
Molecular Weight: 576.108 g/mol
Siedepunkt: 73 °C (10 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD03094451
CAS-Nummer: 330562-42-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR