Tris(dimethylamino)sulphonium difluoro(trimethyl)silicate, technische Qualität

Lieferant: Apollo Scientific
Gefahr

Synonyme: Tris(dimethylamino)sulphur trimethylsilyl difluoride, TAS-F

PC7880-1G PC7880-5G
APOSPC7880-1GEA 169 EUR
APOSPC7880-1G APOSPC7880-5G
Tris(dimethylamino)sulphonium difluoro(trimethyl)silicate, technische Qualität
Tris(dimethylamino)sulphonium difluoro(trimethyl)silicate
Formel: C₃H₉F₂Si·C₆H₁₈N₃S
Molecular Weight: 275.478 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00012170
CAS-Nummer: 59218-87-0
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR