3-Bromo-6-fluoro-2-methoxybenzeneboronic acid 96%
Lieferant: Apollo Scientific
Synonyme:
(3-Bromo-6-fluoro-2-methoxyphenyl)boronic acid, 3-Bromo-6-fluoro-2-methoxyphenylboronic acid
PC7915-1G
PC7915-5G
APOSPC7915-1GEA
85.7
EUR
APOSPC7915-1G
APOSPC7915-5G
3-Bromo-6-fluoro-2-methoxybenzeneboronic acid 96%
3-Bromo-6-fluoro-2-methoxybenzeneboronic acid
Formel:
C₇H₇BBrFO₃ Molecular Weight: 248.842 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09475911 CAS-Nummer: 957120-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...