1-(Perfluorohexyl)octane 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Tridecafluorohex-1-yl)octane, 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluorotetradecane

PC7927-25G PC7927-5G PC7927-1G
APOSPC7927-25GEA 592 EUR
APOSPC7927-25G APOSPC7927-5G APOSPC7927-1G
1-(Perfluorohexyl)octane 97%
1-(Perfluorohexyl)octane
Formel: C₁₄H₁₇F₁₃
Molecular Weight: 432.264 g/mol
Flammpunkt: 100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07784188
CAS-Nummer: 133331-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR