Methyl 2-hydroxy-2-(trifluoromethyl)pent-4-enoate

Lieferant: Apollo Scientific

Synonyme: Methyl 2-hydroxy-2-(trifluoromethyl)-3-vinylpropanoate

PC7944-1G PC7944-5G
APOSPC7944-1GEA 343 EUR
APOSPC7944-1G APOSPC7944-5G
Methyl 2-hydroxy-2-(trifluoromethyl)pent-4-enoate
Methyl 2-hydroxy-2-(trifluoromethyl)pent-4-enoate
Formel: C₇H₉F₃O₃
Molecular Weight: 198,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236694
CAS-Nummer: 117015-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR