1-[3,5-Bis(trifluoromethyl)phenyl]propylamine 97%

Lieferant: Apollo Scientific

Synonyme: 3,5-Bis(trifluoromethyl)-α-ethylbenzylamine

PC7959-1G PC7959-250MG
APOSPC7959-1GEA 361 EUR
APOSPC7959-1G APOSPC7959-250MG
1-[3,5-Bis(trifluoromethyl)phenyl]propylamine 97%
1-[3,5-Bis(trifluoromethyl)phenyl]propylamine
Formel: C₁₁H₁₁F₆N
Molecular Weight: 271,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784287
CAS-Nummer: 685503-45-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR