1-(Trifluorovinyl)cyclohex-1-ene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Trifluoroethenyl)cyclohex-1-ene

PC8008-1G PC8008-5G
APOSPC8008-1GEA 46.4 EUR
APOSPC8008-1G APOSPC8008-5G
1-(Trifluorovinyl)cyclohex-1-ene
1-(Trifluorovinyl)cyclohex-1-ene
Formel: C₈H₉F₃
Molecular Weight: 162,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00156079
CAS-Nummer: 243863-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR