5-Bromo-3-carboxy-2-fluorobenzeneboronic acid
Lieferant: Apollo Scientific
Synonyme:
3-Borono-5-bromo-2-fluorobenzoic acid, 5-Bromo-3-carboxy-2-fluorophenylboronic acid
PC8044-1G
PC8044-5G
APOSPC8044-1GEA
82.8
EUR
APOSPC8044-1G
APOSPC8044-5G
5-Bromo-3-carboxy-2-fluorobenzeneboronic acid
5-Bromo-3-carboxy-2-fluorobenzeneboronic acid
Formel:
C₇H₅BBrFO₄ Molecular Weight: 262,826 g/mol Schmelzpunkt: 237…240 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09800869 CAS-Nummer: 957120-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...