3-Chloro-5-fluorophenetole 97%

Lieferant: Apollo Scientific

PC8161-10G PC8161-1G PC8161-5G
APOSPC8161-10GEA 812 EUR
APOSPC8161-10G APOSPC8161-1G APOSPC8161-5G
3-Chloro-5-fluorophenetole 97%
3-Chloro-5-fluorophenetole
Formel: C₈H₈ClFO
Molecular Weight: 174,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672983
CAS-Nummer: 289039-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR