5-Chloro-2-fluorophenetole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-2-ethoxy-1-fluorobenzene

PC8164-1G PC8164-5G PC8164-25G
APOSPC8164-1GEA 26.1 EUR
APOSPC8164-1G APOSPC8164-5G APOSPC8164-25G
5-Chloro-2-fluorophenetole
5-Chloro-2-fluorophenetole
Formel: C₈H₈ClFO
Molecular Weight: 174,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672980
CAS-Nummer: 289039-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR