1,1,1-Trifluoro-4,4-bis(4,4,4-trifluorobutoxy)butane 97+%

Lieferant: Apollo Scientific

PC8264-1G PC8264-5G
APOSPC8264-1GEA 77.4 EUR
APOSPC8264-1G APOSPC8264-5G
1,1,1-Trifluoro-4,4-bis(4,4,4-trifluorobutoxy)butane 97+%
1,1,1-Trifluoro-4,4-bis(4,4,4-trifluorobutoxy)butane
Formel: C₁₂H₁₇F₉O₂
Molecular Weight: 364.248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08460484
CAS-Nummer: 933600-77-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR