Methyl 2-chloro-4-(trifluoromethyl)cinnamate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-[2-chloro-4-(trifluoromethyl)phenyl]acrylate, Methyl 3-[2-chloro-4-(trifluoromethyl)phenyl]prop-2-enoate

PC8274-100MG PC8274-1G PC8274-250MG
APOSPC8274-100MGEA 208 EUR
APOSPC8274-100MG APOSPC8274-1G APOSPC8274-250MG
Methyl 2-chloro-4-(trifluoromethyl)cinnamate 95+%
Methyl 2-chloro-4-(trifluoromethyl)cinnamate
Formel: C₁₁H₈ClF₃O₂
Molecular Weight: 264,628 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01313366
CAS-Nummer: 243977-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR