4-(Bromomethyl)-N-(4-fluorophenyl)benzenesulphonamide

Lieferant: Apollo Scientific
Achtung

Synonyme: ((4-(Bromomethyl)phenyl)sulphonyl)(4-fluorophenyl)amine

PC8281-1G PC8281-250MG PC8281-5G
APOSPC8281-1GEA 139 EUR
APOSPC8281-1G APOSPC8281-250MG APOSPC8281-5G
4-(Bromomethyl)-N-(4-fluorophenyl)benzenesulphonamide
4-(Bromomethyl)-N-(4-fluorophenyl)benzenesulphonamide
Formel: C₁₃H₁₁BrFNO₂S
Molecular Weight: 344,199 g/mol
Schmelzpunkt: 141…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00169942
CAS-Nummer: 260969-08-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR