Bromo(difluoro)acetyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-2,2-difluoroethanoyl bromide

PC8341-1G PC8341-25G PC8341-5G
APOSPC8341-1GEA 72.5 EUR
APOSPC8341-1G APOSPC8341-25G APOSPC8341-5G
Bromo(difluoro)acetyl bromide 97%
Bromo(difluoro)acetyl bromide
Formel: C₂Br₂F₂O
Molecular Weight: 237.826 g/mol
Siedepunkt: 70…79 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00153688
CAS-Nummer: 1796-12-9
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR