3,3-Difluoroallyl acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,3-Difluoroprop-2-en-1-yl acetate

PC8369-1G PC8369-5G
APOSPC8369-1GEA 263 EUR
APOSPC8369-1G APOSPC8369-5G
3,3-Difluoroallyl acetate
3,3-Difluoroallyl acetate
Formel: C₅H₆F₂O₂
Molecular Weight: 136,097 g/mol
Siedepunkt: 59 °C (150 mm Hg)
Lagertemperatur: Kühlschrank
MDL: MFCD16170466
CAS-Nummer: 1597-40-6
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR