4-Bromo-2-nitro-6-(trifluoromethoxy)aniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-bromo-3-(trifluoromethoxy)nitrobenzene
PC8373-1G
PC8373-250MG
PC8373-5G
APOSPC8373-1GEA
52.2
EUR
APOSPC8373-1G
APOSPC8373-250MG
APOSPC8373-5G
4-Bromo-2-nitro-6-(trifluoromethoxy)aniline
4-Bromo-2-nitro-6-(trifluoromethoxy)aniline
Formel:
C₇H₄BrF₃N₂O₃ Molecular Weight: 301,017 g/mol Schmelzpunkt: 125…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16628052 CAS-Nummer: 1257535-31-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...