5-Bromo-3-(trifluoromethoxy)benzene-1,2-diamine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8378-1G
PC8378-250MG
APOSPC8378-1GEA
453
EUR
APOSPC8378-1G
APOSPC8378-250MG
5-Bromo-3-(trifluoromethoxy)benzene-1,2-diamine 97%
5-Bromo-3-(trifluoromethoxy)benzene-1,2-diamine
Formel:
C₇H₆BrF₃N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD16628053 CAS-Nummer: 1257535-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...