2-Fluoro-5-(2-methylallyl)toluene
Lieferant: Apollo Scientific
Synonyme:
3-(4-Fluoro-3-methylphenyl)-2-methylprop-1-ene
PC8390-1G
PC8390-5G
APOSPC8390-1GEA
239
EUR
APOSPC8390-1G
APOSPC8390-5G
2-Fluoro-5-(2-methylallyl)toluene
2-Fluoro-5-(2-methylallyl)toluene
Formel:
C₁₁H₁₃F Molecular Weight: 164.219 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07775007 CAS-Nummer: 869493-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...