Ethyl 3-amino-3-[3-(trifluoromethyl)anilino]prop-2-en-1-oate hydrochloride

Lieferant: Apollo Scientific

PC8466-1G PC8466-500MG
APOSPC8466-1GEA 590 EUR
APOSPC8466-1G APOSPC8466-500MG
Ethyl 3-amino-3-[3-(trifluoromethyl)anilino]prop-2-en-1-oate hydrochloride
Ethyl 3-amino-3-[3-(trifluoromethyl)anilino]prop-2-en-1-oate hydrochloride
Formel: C₁₂H₁₃F₃N₂O₂·ClH
Molecular Weight: 310,7 g/mol
Schmelzpunkt: 158…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00243964
CAS-Nummer: 318469-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR