4-(Trifluoromethylthio)phenacyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-4'-(trifluoromethylthio)acetophenone

PC8506-1G PC8506-250MG
APOSPC8506-1GEA 833 EUR
APOSPC8506-1G APOSPC8506-250MG
4-(Trifluoromethylthio)phenacyl bromide
4-(Trifluoromethylthio)phenacyl bromide
Formel: C₉H₆BrF₃OS
Molecular Weight: 299.108 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03094514
CAS-Nummer: 2003-11-4
UN: 3261
ADR: 8 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR