2-[4-(Chloromethyl)phenyl]-5-(trifluoromethyl)pyridine 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
6-[4-(Chloromethyl)phenyl]-α,α,α-trifluoro-3-picoline, 4-[5-(Trifluoromethyl)pyridin-2-yl]benzyl chloride
PC8552-1G
PC8552-250MG
PC8552-100MG
APOSPC8552-1GEA
0
EUR
APOSPC8552-1G
APOSPC8552-250MG
APOSPC8552-100MG
2-[4-(Chloromethyl)phenyl]-5-(trifluoromethyl)pyridine 95%
2-[4-(Chloromethyl)phenyl]-5-(trifluoromethyl)pyridine
Formel:
C₁₃H₉ClF₃N Molecular Weight: 271.665 g/mol Schmelzpunkt: 77…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09817479 CAS-Nummer: 613239-76-2 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...